missing translation for 'onlineSavingsMsg'
Learn More

n-Heptylamine, 99+%

Product Code. 15179193 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15179193 500 mL Glass bottle 500mL
10071690 25 mL Glass bottle 25mL
10081690 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15179193 Supplier Thermo Scientific Acros Supplier No. 155465000
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 111-68-2 | C7H17N | 115.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-68-2
Molecular Formula C7H17N
Molecular Weight (g/mol) 115.22
MDL Number MFCD00008244
InChI Key WJYIASZWHGOTOU-UHFFFAOYSA-N
Synonym heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681
PubChem CID 8127
IUPAC Name heptan-1-amine
SMILES CCCCCCCN

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -23.0°C
Color Colorless to Yellow
Density 0.7740g/mL
Boiling Point 153.0°C to 156.0°C
Flash Point 44°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3(CH2)6NH2
Refractive Index 1.4233 to 1.4253
Quantity 500 mL
Beilstein 04,193
Specific Gravity 0.774
Solubility Information Solubility in water: 100g/L water (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 115.22
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material n-Heptylamine, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.