missing translation for 'onlineSavingsMsg'
Learn More

n-Hexadecyl palmitate, 95%

Product Code. 10633415 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10633415 250 g 250g
10794565 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10633415 Supplier Thermo Scientific Acros Supplier No. 428222500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 540-10-3 | C32H64O2 | 480.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-10-3
Molecular Formula C32H64O2
Molecular Weight (g/mol) 480.86
MDL Number MFCD00053739
InChI Key PXDJXZJSCPSGGI-UHFFFAOYSA-N
Synonym cetyl palmitate, hexadecyl palmitate, palmityl palmitate, cetin, hexadecanoic acid, hexadecyl ester, standamul 1616, hexadecanoic acid hexadecyl ester, hexadecanyl hexadecanoate, n-hexadecyl hexadecanoate, palmitic acid palmityl ester
PubChem CID 10889
ChEBI CHEBI:75584
IUPAC Name hexadecyl hexadecanoate
SMILES CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC

Specifications

Melting Point 52°C to 56°C
Boiling Point 360°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Linear Formula CH3(CH2)14COO(CH2)15CH3
Quantity 250 g
Merck Index 15, 2030
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 480.86
Percent Purity 95%
Chemical Name or Material n-Hexadecyl palmitate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.