missing translation for 'onlineSavingsMsg'
Learn More

(n-Hexadecyl)tri-n-butylphosphonium bromide, 98+%

Product Code. 11394564
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394564 100 g 100g
11374564 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394564 Supplier Thermo Scientific Alfa Aesar Supplier No. L01335.22
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14937-45-2 | C28H60BrP | 507.67 g/mol

(n-Hexadecyl)tri-n-butylphosphonium bromide is used as Phase-transfer-catalyst. It is a eco friendly highly effective water treatment agent with low foaming and toxic-free.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(n-Hexadecyl)tri-n-butylphosphonium bromide is used as Phase-transfer-catalyst. It is a eco friendly highly effective water treatment agent with low foaming and toxic-free.

Solubility
Soluble in methanol.

Notes
Hygroscopic. Protect from carbondioxide. Store away from moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14937-45-2
Molecular Formula C28H60BrP
Molecular Weight (g/mol) 507.67
MDL Number MFCD00011775
InChI Key RYVBINGWVJJDPU-UHFFFAOYSA-M
Synonym tributylhexadecylphosphonium bromide, tributyl hexadecyl phosphonium bromide, cetyltributylphosphonium bromide, tributyl hexadecyl phosphanium bromide, phosphonium, tributylhexadecyl-, bromide, hexadecyltributylphosphonium bromide, phosphonium, tributylhexadecyl-, bromide 1:1, hexadecyltri-n-butylphosphonium bromide, n-hexadecyl tri-n-butylphosphonium bromide, tbhdpb
PubChem CID 84716
SMILES [Br-].CCCCCCCCCCCCCCCC[P+](CCCC)(CCCC)CCCC

Specifications

Melting Point 56°C to 62°C
Quantity 100 g
Beilstein 4167404
Sensitivity Hygroscopic
Solubility Information Soluble in methanol.
IUPAC Name tributyl(hexadecyl)phosphanium;bromide
Formula Weight 507.67
Percent Purity ≥98%
Chemical Name or Material (n-Hexadecyl)tri-n-butylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.