missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Hexadecyltrimethyl-d42-ammonium p-Toluene-d7-sulfonate, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Hexadecyltrimethylammonium-D42 Tosylate-D7 |
| Molecular Formula | C19 D42 N . C7 D7 O3 S |
| Quantity | 0.01 g |
| Synonym | N,N,N-Tri(methyl-d3)-1-hexadecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-d33-aminium 4-(methyl-d3)benzenesulfonate-2,3,5,6-d4, Hexadecyltrimethylammonium-d42 p-toluene-d7-sulfonate, Cetrimonium-d42 tosylate-d7, Hexadecyltrimethylammonium-D42 tosylate-D7 |
| InChI Formula | InChI=1 S/C19H42N.C7H8O3S/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;1-6-2-4-7(5-3-6)11(8,9)10/h5-19H2,1-4H3;2-5 H,1H3,(H,8,9,10)/q+1;/p-1/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2;1D3,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1C([2 H])([2 H])[2 H])S(=O)(=O)[O-].[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[N+](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-(trideuteriomethyl)benzenesulfonate;tris(trideuteriomethyl)-(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-tritriacontadeuteriohexadecyl)azanium |
| Molecular Weight (g/mol) | 505.039 |
| Formula Weight | 504.651 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction