missing translation for 'onlineSavingsMsg'
Learn More

n-Hexanal dimethylacetal, 98+%, Thermo Scientific™

Product Code. 10712294 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10712294 5 g Glass bottle 5g
10722294 25 g Glass bottle 25g
10454135 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10712294 Supplier Thermo Scientific Acros Supplier No. 393560050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1599-47-9
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00036508
InChI Key ZQUJUCJLDXTKKW-UHFFFAOYSA-N
PubChem CID 74137
IUPAC Name 1,1-dimethoxyhexane
SMILES CCCCCC(OC)OC

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless
Density 0.8470g/mL
Boiling Point 145°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4CH(OCH3)2
Refractive Index 1.4050 to 1.4070 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.847
Synonym hexanal dimethyl acetal, hexane, 1,1-dimethoxy, aldehyde c-6 dimethyl acetal, acmc-20akrj, 1,1-dimethoxy-hexane, hexane,1,1-dimethoxy, hexaldehyde dimethyl acetal, 1,1-dimethoxyhexane
Solubility Information Solubility in water: practically insoluble
Formula Weight 146.23
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material n-Hexanal dimethylacetal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • n-Hexanal dimethylacetal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.