missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, 96+%, Extra Dry, AcroSeal™

Product Code. 10061871 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10061871 100 mL 100mL
10292412 1 L 1 L
10359233 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 10061871 Supplier Thermo Scientific Acros Supplier No. 326921000
  • ONLINE EXCLUSIVE PRICE
    32.15 €
    31.20 € / 100mL
    Save 0.95 €
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 110-54-3 | C6H14 | 86.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95.0°C
CAS Min % 96.0
Color Colorless
CAS Max % 100.0
Density 0.6590g/mL
Quantity 100 mL
Flash Point −22°C
Packaging AcroSeal™ Glass Bottle
Physical Form Liquid
Refractive Index 1.3748 to 1.3810
Residue after Evaporation 0.0005 % max.
Assay Percent Range 96+%
Specific Gravity 0.659
Linear Formula CH3(CH2)4CH3
Viscosity 0.31 mPa.s (20°C)
Water 0.002% Max.
Beilstein 01, 142
Merck Index 15, 4732
Boiling Point 69.0°C
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Formula Weight 86.18
Percent Purity ≥96%
Grade Extra Dry
Chemical Name or Material n-Hexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Hexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.