Learn More
N-Hexane, Certified AR for Analysis, Fisher Chemical™
C6H14, CAS Number-110-54-3, hexanes, hexyl hydride, hexan, hexanen, gettysolve-b, heksan, dipropyl, esani, n-hexane, skellysolve b, 2.5L, 69 deg.C, CHEBI:29021, Colorless, 86.18g/mol, VLKZOEOYAKHREP-UHFFFAOYSA-N, -95 deg.C, 2179311, 86.178, Plastic Coated Glass bottle, Liquid, 8058, CCCCCC, 2.97
Brand: Fisher Chemical H/0421/PB17
214.61 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Code : 22
Additional Details : CAS Number : 110-54-3 Weight : 1.64750kg Transport : UN number : 1208 Chem class : 3 Pack group : II
Description
C6H14, CAS Number-110-54-3, hexanes, hexyl hydride, hexan, hexanen, gettysolve-b, heksan, dipropyl, esani, n-hexane, skellysolve b, 2.5L, 69 deg.C, CHEBI:29021, Colorless, 86.18g/mol, VLKZOEOYAKHREP-UHFFFAOYSA-N, -95 deg.C, 2179311, 86.178, Plastic Coated Glass bottle, Liquid, 8058, CCCCCC, 2.97
Specifications
-95°C | |
Colorless | |
2.5 L | |
Liquid | |
MFCD02179311 | |
0.31 mPaS at 20°C | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
hexane | |
8058 | |
86.18g/mol |
110-54-3 | |
69°C | |
Plastic-coated Glass Bottle | |
C6H14 | |
160mbar at 20°C | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
CCCCCC | |
86.18 | |
CHEBI:29021 |
Safety and Handling
- n-Hexane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P273-Avoid release to the environment.