Product Code. 16695893
Click to view available options
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
unitSize
16695893
7 L
7 L
16685893
2.5 L
2.50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code.
16695893
Supplier
Burdick & Jackson BioSyn
Supplier No.
657477LRC
Estimated Shipment: 10-11-2026Log in to see stock.
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Chemical Identifiers
CAS
110-54-3
Molecular Formula
C6H14
Molecular Weight (g/mol)
86.18
MDL Number
MFCD02179311
InChI Key
VLKZOEOYAKHREP-UHFFFAOYSA-N
IUPAC Name
hexane
SMILES
CCCCCC
Specifications
Melting Point
-95°C
Color
Colorless
CAS Max %
1
Density
0.660 - 0.668 g/cm3 (20°C)
Quantity
7 L
Flash Point
-26°C
Boiling Point
66°C to 69°C (1013 hPa)
Chemical Name or Material
n-Hexane
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?