missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, Puriss. p.a., Reagent ACS, Reagent European Pharmacopoeia, Honeywell Burdick & Jackson™

Product Code. 16665903
Change view
Click to view available options
:
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16665903 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 16665903 Supplier Honeywell Burdick & Jackson Supplier No. 65801200L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Specifications

Melting Point -95°C
CAS 110-54-3
Color Colorless
CAS Max % 1
Density 0.660 - 0.668 g/cm3 (20°C)
Quantity 200 L
Flash Point -26°C
Molecular Formula C6H14
Linear Formula CH3(CH2)4CH3
MDL Number MFCD02179311
Boiling Point 66°C to 69°C (1013 hPa)
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES CCCCCC
IUPAC Name hexane
Molecular Weight (g/mol) 86.18
Grade ACS
Chemical Name or Material n-Hexane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.