missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Hexanoyl-d11-glycine, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Hexanoyl-d11 Glycine |
| Molecular Formula | C8 D11 H4 N O3 |
| Quantity | 0.05 g |
| Synonym | N-(1-Oxohexyl)glycine-d11, Caproylglycine-d11, N-Caproylglycine-d11, NSC 224460-d11, n-Hexanoylglycine-d11, N-Hexanoyl-d11-glycine, N-(1-Oxohexyl-2,2,3,3,4,4,5,5,6,6,6-d11)glycine, N-Hexanoyl-d11-glycine, N-Hexanoylglycine-D11 (hexanoyl-D11) |
| InChI Formula | InChI=1 S/C8H15NO3/c1-2-3-4-5-7(10)9-6-8(11)12/h2-6H2,1H3,(H,9,10)(H,11,12)/i1D3,2D2,3D2,4D2,5D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)NCC(=O)O |
| IUPAC Name | 2-(2,2,3,3,4,4,5,5,6,6,6-undecadeuteriohexanoylamino)acetic acid |
| Molecular Weight (g/mol) | 184.28 |
| Formula Weight | 184.1742 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction