missing translation for 'onlineSavingsMsg'
Learn More

n-Hexyl acrylate, 95%, stab. with hydroquinone, Thermo Scientific Chemicals

Product Code. 11352347
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352347 100 g 100g
11342347 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352347 Supplier Thermo Scientific Alfa Aesar Supplier No. L09905.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2499-95-8
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00048881
InChI Key LNMQRPPRQDGUDR-UHFFFAOYSA-N
Synonym hexyl acrylate, n-hexyl acrylate, hexyl 2-propenoate, 2-propenoic acid, hexyl ester, n-hexylacrylate, ageflex n-ha, acrylic acid, hexyl ester, ccris 7038, acrylic acid-n-hexyl ester, hexylacrylate
PubChem CID 17259
IUPAC Name hexyl prop-2-enoate
SMILES CCCCCCOC(=O)C=C

Specifications

Melting Point -45°C
Density 0.89
Boiling Point 88°C to 89°C (24 mmHg)
Flash Point 68°C (154°F)
Linear Formula H2C=CHCO2(CH2)5CH3
Refractive Index 1.428
Quantity 100 g
UN Number UN3082
Beilstein 1757327
Formula Weight 156.22
Percent Purity 95%
Chemical Name or Material n-Hexyl acrylate, stabilized with hydroquinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.