missing translation for 'onlineSavingsMsg'
Learn More

n-Hexylphosphonic acid

Product Code. 11399598
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399598 10 g 10g
11389598 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399598 Supplier Thermo Scientific Alfa Aesar Supplier No. 044121.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4721-24-8 | C6H15O3P | 166.157 g/mol

n-Hexylphosphonic acid is used in the manufacturing of nanoparticles such as quantum dots, nano-metals and nano-ceramics. It is also used for coating of many materials, including nanoparticles by condensed hydrophobic monolayers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4721-24-8
Molecular Formula C6H15O3P
Molecular Weight (g/mol) 166.157
MDL Number MFCD00013961
InChI Key GJWAEWLHSDGBGG-UHFFFAOYSA-N
Synonym n-hexylphosphonic acid, phosphonic acid, p-hexyl, hexylphosphonicacid, p-hexyl-phosphonic acid, hexylphosphonic acid
PubChem CID 312552
IUPAC Name hexylphosphonic acid
SMILES CCCCCCP(=O)(O)O

Specifications

Melting Point 108°C to 110°C
Color White
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 166.16
Physical Form Powder
Chemical Name or Material n-Hexylphosphonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.