missing translation for 'onlineSavingsMsg'
Learn More

n'-Hydroxy-2-phenoxyethanimidamide, ≥97%, Thermo Scientific™

Product Code. 10037502
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10037502 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10037502 Supplier Thermo Scientific Maybridge Supplier No. SPB02646EA
 Restricted Item
Estimated Shipment: 19-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 90007-06-0
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H10N2O2
MDL Number MFCD02090467
Quantity 10 g
Synonym 1z-n'-hydroxy-2-phenoxyethanimidamide, n'-hydroxy-2-phenoxyacetimidamide, 1-hydroxyimino-2-phenoxyethylamine, z-n'-hydroxy-2-phenoxyacetimidamide, z-n'-hydroxy-2-phenoxyethenimidamide
InChI Key BIIYCNCZOGOUJP-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)OCC(=NO)N
IUPAC Name N'-hydroxy-2-phenoxyethanimidamide
Molecular Weight (g/mol) 166.18
PubChem CID 9580876
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material N'-Hydroxy-2-phenoxyethanimidamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.