missing translation for 'onlineSavingsMsg'
Learn More

n-Hydroxy-butyramidine, ≥97%, Thermo Scientific™

Product Code. 10632025
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10632025 1 g Amber Glass Bottle 1g
10533423 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10632025 Supplier Thermo Scientific Maybridge Supplier No. MO07267DA
 Restricted Item
Estimated Shipment: 03-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 27620-10-6
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.137
MDL Number MFCD06200896
InChI Key OPENCMFJZQABIY-UHFFFAOYSA-N
Synonym n-hydroxybutyramidine, 1z-n'-hydroxybutanimidamide, n-hydroxybutyrimidamide, z-n'-hydroxybutyrimidamid e, butanimidamide, n-hydroxy, z-n'-hydroxybutimidamide, butanimidamide,n-hydroxy, e-n'-hydroxybutanimidamide, 1-1h-benzo d imidazol-2-yl-3-methyl-4-oxo-1,4-di
PubChem CID 9582833
IUPAC Name N'-hydroxybutanimidamide
SMILES CCCC(=NO)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 102.14
Percent Purity 97%
Chemical Name or Material N-hydroxy-butyramidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.