missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxybenzenecarboximidamide, 97%, Thermo Scientific™

Product Code. 10551122
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10551122 1 g Amber Glass Bottle 1g
10457432 10 g Amber Glass Bottle 10g
10557474 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10551122 Supplier Thermo Scientific Maybridge Supplier No. RF01827DA
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 613-92-3
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00031485 MFCD00474011
InChI Key MXOQNVMDKHLYCZ-UHFFFAOYSA-N
Synonym benzamidoxime, benzamide oxime, n'-hydroxybenzimidamide, benzohydroxamamide, n-hydroxybenzamidine, phenylhydroxamidine, n-hydroxybenzenecarboximidamide, ccris 2953, n'-hydroxybenzamidine, benzamidoxim
PubChem CID 7259353
ChEBI CHEBI:83354
SMILES N\C(=N/O)C1=CC=CC=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula C6H5C(NH2)=NOH
Quantity 1 g
IUPAC Name N'-hydroxybenzenecarboximidamide
Formula Weight 136.15
Percent Purity 97%
Chemical Name or Material N'-Hydroxybenzenecarboximidamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.