missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxyethyl-N-methyl-tryptamine, TRC

Product Code. 30010737
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010737 100 mg 100mg
30006489 10 mg 10mg
30006889 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010737 Supplier Toronto Research Chemicals Supplier No. TRCH727265100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101264-45-3
Molecular Formula C13 H18 N2 O
Molecular Weight (g/mol) 218.29
Synonym 2-[[2-(1H-Indol-3-yl)ethyl]methylamino]ethanol;2-[(2-Indol-3-yl-ethyl)-methyl-amino]-ethanol;2-[[2-(1H-Indol-3-yl)ethyl](methyl)amino]ethan-1-ol
IUPAC Name 2-[2-(1H-indol-3-yl)ethyl-methyl-amino]ethanol
SMILES CN(CCO)CCc1c[nH]c2ccccc12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H18N2O/c1-15(8-9-16)7-6-11-10-14-13-5-3-2-4-12(11)13/h2-5,10,14,16H,6-9H2,1H3
Formula Weight 218.1419
Chemical Name or Material N-Hydroxyethyl-N-methyl-tryptamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.