missing translation for 'onlineSavingsMsg'
Learn More

N-(Hydroxymethyl)phthalimide, 97%

Product Code. 11409207
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11409207

missing translation for 'mfr': Thermo Scientific Alfa Aesar B21292.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118-29-6 | C9H7NO3 | 177.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-29-6
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.159
MDL Number MFCD00005899
InChI Key MNSGOOCAMMSKGI-UHFFFAOYSA-N
Synonym n-hydroxymethyl phthalimide, n-hydroxymethylphthalimide, 2-hydroxymethyl isoindoline-1,3-dione, phthalimidomethanol, phthalimidomethyl alcohol, n-methylolphthalimide, oxymethyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxymethyl, hydroxymethylphthalimide, methanol, phthalimido
PubChem CID 8354
ChEBI CHEBI:38816
IUPAC Name 2-(hydroxymethyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CO

Specifications

Melting Point 140°C to 147°C
Quantity 25 g
Beilstein 140946
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material N-(Hydroxymethyl)phthalimide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.