missing translation for 'onlineSavingsMsg'
Learn More

N-(Hydroxymethyl)phthalimide, 97%

Product Code. 10606702 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10606702 25 g Glass bottle 25g
10245400 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10606702 Supplier Thermo Scientific Acros Supplier No. 121490250
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 118-29-6 | C9H7NO3 | 177.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-29-6
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.16
MDL Number MFCD00005899
InChI Key MNSGOOCAMMSKGI-UHFFFAOYSA-N
Synonym n-hydroxymethyl phthalimide, n-hydroxymethylphthalimide, 2-hydroxymethyl isoindoline-1,3-dione, phthalimidomethanol, phthalimidomethyl alcohol, n-methylolphthalimide, oxymethyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxymethyl, hydroxymethylphthalimide, methanol, phthalimido
PubChem CID 8354
ChEBI CHEBI:38816
IUPAC Name 2-(hydroxymethyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 140°C to 145°C
Color White
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 21, 475
Fieser 01,484; 08,253
Formula Weight 177.16
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material N-(Hydroxymethyl)phthalimide, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.