missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxyphthalimide, 98%

Product Code. 11471528
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471528 500 g 500g
11461528 100 g 100g
11481528 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Código de producto. 11471528 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor A13862.36
 Restricted Item
Estimated Shipment: 25-11-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 524-38-9 | C8H5NO3 | 163.13 g/mol

Aerobic oxidation of various alcohols has been accomplished by using a new catalytic system, N-hydroxyphthalimide (NHPI) combined with Co (acac). A practical catalytic method to convert alkylbenzenes into the corresponding carboxylic acids under atmospheric dioxygen at ambient temperature using a combined catalytic system consisting of N-hydroxyphthalimide (NHPI) and Co (OAc) 2 was developed. Novel catalysis by N-hydroxyphthalimide in the oxidation of organic substrates by molecular oxygen was described. Free radical functionalization of organic compounds catalyzed by N-hydroxyphthalimide. Purely organic and catalytic systems of anthraquinones and N-hydroxyphthalimide efficiently promote oxygenation of hydrocarbons with dioxygen under mild conditions. Hydroxylation of polycyclic alkanes with molecular oxygen catalyzed by N-hydroxyphthalimide (NHPI) combined with was transition metal salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aerobic oxidation of various alcohols has been accomplished by using a new catalytic system, N-hydroxyphthalimide (NHPI) combined with Co (acac). A practical catalytic method to convert alkylbenzenes into the corresponding carboxylic acids under atmospheric dioxygen at ambient temperature using a combined catalytic system consisting of N-hydroxyphthalimide (NHPI) and Co (OAc) 2 was developed. Novel catalysis by N-hydroxyphthalimide in the oxidation of organic substrates by molecular oxygen was described. Free radical functionalization of organic compounds catalyzed by N-hydroxyphthalimide. Purely organic and catalytic systems of anthraquinones and N-hydroxyphthalimide efficiently promote oxygenation of hydrocarbons with dioxygen under mild conditions. Hydroxylation of polycyclic alkanes with molecular oxygen catalyzed by N-hydroxyphthalimide (NHPI) combined with was transition metal salts.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 524-38-9
Molecular Formula C8H5NO3
Molecular Weight (g/mol) 163.13
MDL Number MFCD00005891
InChI Key CFMZSMGAMPBRBE-UHFFFAOYSA-N
Synonym n-hydroxyphthalimide, 2-hydroxyisoindoline-1,3-dione, 2-hydroxy-1h-isoindole-1,3 2h-dione, nhpi, 2-hydroxyphthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxy, phthalimide, n-hydroxy, phthaloxime, unii-bxi99m81x0, 2-hydroxyisoindolin-1,3-dione
PubChem CID 10665
IUPAC Name 2-hydroxyisoindole-1,3-dione
SMILES ON1C(=O)C2=CC=CC=C2C1=O

Especificaciones

Melting Point ∼233°C (decomposition)
Quantity 500 g
Beilstein 131208
Merck Index 14,4838
Solubility Information Slightly soluble in water.
Formula Weight 163.13
Percent Purity ≥98%
Chemical Name or Material N-Hydroxyphthalimide

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.