missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxysulfosuccinimide sodium salt, 97%

Product Code. 15489547
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15489547 250 mg 250mg
15469547 1 g 1g
15479547 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15489547 Supplier Thermo Scientific Alfa Aesar Supplier No. H52795.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106627-54-7 | C4H4NNaO6S | 217.127 g/mol

N-Hydroxysulfosuccinimide sodium salt is used as crosslinking agents for preparing hydrophilic active esters. It is also used to activate carboxylic acids to the more reactive succinimidyl esters for coupling, biotinylation and conjugation with fluorophores. Further, it acts as a hydrophilic ligand and also used in covalently binding proteins to carboxyl containing beads, resins, other nanoparticles. It is involved in the conversion of carboxyl groups to primary amine-reactive sulfo-NHS esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Hydroxysulfosuccinimide sodium salt is used as crosslinking agents for preparing hydrophilic active esters. It is also used to activate carboxylic acids to the more reactive succinimidyl esters for coupling, biotinylation and conjugation with fluorophores. Further, it acts as a hydrophilic ligand and also used in covalently binding proteins to carboxyl containing beads, resins, other nanoparticles. It is involved in the conversion of carboxyl groups to primary amine-reactive sulfo-NHS esters.

Solubility
Soluble in methanol and water.

Notes
Store in a cool place. Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106627-54-7
Molecular Formula C4H4NNaO6S
Molecular Weight (g/mol) 217.127
MDL Number MFCD00043100
InChI Key RPENMORRBUTCPR-UHFFFAOYSA-M
Synonym n-hydroxysulfosuccinimide sodium salt, sulfo-nhs, sodium 1-hydroxy-2,5-dioxopyrrolidine-3-sulfonate, n-hydroxysulfosuccinimidesodiumsalt, 3-pyrrolidinesulfonic acid, 1-hydroxy-2,5-dioxo-, monosodium salt, sodium 1-hydroxy-2,5-dioxo-pyrrolidine-3-sulfonate, n-hydroxusulfosuccinimide sodium, n-hydroxy-2-sodiosulfo succinimide, n-hydroxysulfosuccinimide,sodium salt, n-hydroxusulfosuccinimide sodium salt
PubChem CID 3520574
IUPAC Name sodium;1-hydroxy-2,5-dioxopyrrolidine-3-sulfonate
SMILES C1C(C(=O)N(C1=O)O)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point 250°C
Quantity 250 mg
Beilstein 6359129
Solubility Information Soluble in methanol and water.
Formula Weight 217.13
Percent Purity 97%
Chemical Name or Material N-Hydroxysulfosuccinimide sodium salt
compliance-icons
Product Identifier
  • N-Hydroxysulfosuccinimide sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : N-Hydroxysulfosuccinimide sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.