missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Isopropylacrylamide, 99%, pure, stabilized

Product Code. 10730102 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10730102 100 g 100g
10537942 25 g 25g
10239700 500 g 500g
15284724 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10730102 Supplier Thermo Scientific Chemicals Supplier No. 412781000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2210-25-5 | C6H11NO | 113.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2210-25-5
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00041913
InChI Key QNILTEGFHQSKFF-UHFFFAOYSA-N
Synonym n-isopropylacrylamide, nipam, n-iso-propylacrylamide, 2-propenamide, n-1-methylethyl, isopropyl acrylamide, acrylamide, n-isopropyl, n-isopropyl acrylamide, n-1-methylethyl-2-propenamide, isopropylamid kyseliny akrylove, unii-b7gff17l9u
PubChem CID 16637
IUPAC Name N-propan-2-ylprop-2-enamide
SMILES CC(C)NC(=O)C=C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 62.0°C to 66.0°C
Color White to Yellow
Boiling Point 89.0°C to 92.0°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH2CHCONHCH(CH3)2
Quantity 100 g
Solubility Information Solubility in water: soluble
Stabilizer 500ppm (ca.) MEHQ
Formula Weight 113.16
Percent Purity 99%
Grade Pure
Physical Form Crystalline Powder or Crystals
Chemical Name or Material N-Isopropylacrylamide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • N-Isopropylacrylamide, stabilized
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.