missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylcyclohexylamine, 99%

Product Code. 11992051 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11992051 25 g Glass bottle 25g
10062190 5 g Glass bottle 5g
10226180 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11992051 Supplier Thermo Scientific Acros Supplier No. 171760250
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1195-42-2 | C9H19N | 141.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1195-42-2
Molecular Formula C9H19N
Molecular Weight (g/mol) 141.26
MDL Number MFCD00003831
InChI Key UYYCVBASZNFFRX-UHFFFAOYSA-N
Synonym n-isopropylcyclohexylamine, cyclohexylisopropylamine, n-isopropylcyclohexanamine, cyclohexylamine, n-isopropyl, isopropylcyclohexylamine, cyclohexanamine, n-1-methylethyl, n-cyclohexylisopropylamine, n-cyclohexyl-n-isopropylamine, n-propan-2-yl cyclohexanamine, n-isopropyl cyclohexylamine
PubChem CID 62386
IUPAC Name N-propan-2-ylcyclohexanamine
SMILES CC(C)NC1CCCCC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8500g/mL
Boiling Point 175°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H11NHCH(CH3)2
Refractive Index 1.4470 to 1.449
Quantity 25 g
Specific Gravity 0.85
Formula Weight 141.26
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Isopropylcyclohexylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Isopropylcyclohexylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.