missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylpropylamine, 96%, Thermo Scientific Chemicals

Product Code. 11429390
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429390 1 g 1g
11419390 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429390 Supplier Thermo Scientific Alfa Aesar Supplier No. H50256.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

N-Isopropylpropylamine is used as an active pharmaceutical ingredients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Isopropylpropylamine is used as an active pharmaceutical ingredients.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21968-17-2
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.193
MDL Number MFCD00027095
InChI Key VLSTXUUYLIALPB-UHFFFAOYSA-N
Synonym n-isopropylpropylamine, propan-2-yl propyl amine, n-propyl-n-isopropylamine, unii-k1p2fy26ro, k1p2fy26ro, 1-propanamine, n-1-methylethyl, propyl-isopropylamine, acmc-20ak2r, n-isopropyl-n-propylamine
PubChem CID 89119
IUPAC Name N-propan-2-ylpropan-1-amine
SMILES CCCNC(C)C

Specifications

Linear Formula CH3CH2CH2NHCH(CH3)2
Quantity 1 g
UN Number UN2733
Formula Weight 101.19
Percent Purity 96%
Chemical Name or Material N-Isopropylpropylamine
compliance-icons compliance-icons
Product Identifier
  • N-isopropylpropylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: N-Isopropylpropylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.