missing translation for 'onlineSavingsMsg'
Learn More

N-Lauroyl Sarcosine Sodium Salt MP Biomedicals

Product Code. 11441821 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441821 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11441821 Supplier MP Biomedicals Supplier No. 0219028950
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 137-16-6
Quantity 50 g
Molecular Formula C15H28NNaO3
MDL Number MFCD00042728
Synonym Sarkosyl NL-97, Sodium N-Lauroyl Sarcosinate
InChI Key KSAVQLQVUXSOCR-UHFFFAOYSA-M
SMILES [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O
IUPAC Name sodium 2-(N-methyldodecanamido)acetate
Molecular Weight (g/mol) 293.38
PubChem CID 23668817
Formula Weight 293.4
Chemical Name or Material N-Lauroyl Sarcosine Sodium Salt
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N-Lauroylsarcosine Sodium Salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P320-Specific treatment is urgent (see on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: N-Lauroylsarcosine Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.