missing translation for 'onlineSavingsMsg'
Learn More

N-Lauroylsarcosine, MP Biomedicals™

Product Code. 11496471 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496471 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11496471 Supplier MP Biomedicals Supplier No. 0219400990
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 137-16-6
Molecular Formula C15H28NNaO3
MDL Number MFCD00042728
Quantity 500 g
Synonym sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
InChI Key KSAVQLQVUXSOCR-UHFFFAOYSA-M
SMILES [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O
IUPAC Name sodium 2-(N-methyldodecanamido)acetate
Molecular Weight (g/mol) 293.38
PubChem CID 23668817
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.