missing translation for 'onlineSavingsMsg'
Learn More

N-Lauroylsarcosine sodium salt, 95%

Product Code. 15412889
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15412889 50 g 50g
15402889 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15412889 Supplier Thermo Scientific Alfa Aesar Supplier No. J60040.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 137-16-6 | C15H28NNaO3 | 293.38 g/mol

N-Lauroylsarcosine is used for solubilization and separation of membrane proteins and glycoprotein's; reported to inhibit hexokinase. It is useful in concentrated salt solutions used in the cell lysis step during RNA purification (helps avoid excessive foaming). It has been used to indicate paramagnetic anisotropy sign change in micelle mesophage. Inhibits bacterial flora of human saliva/gut at 0.25% as well as acting as a fungi static agent in aqueous dispersion (1%).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Lauroylsarcosine is used for solubilization and separation of membrane proteins and glycoprotein′s; reported to inhibit hexokinase. It is useful in concentrated salt solutions used in the cell lysis step during RNA purification (helps avoid excessive foaming). It has been used to indicate paramagnetic anisotropy sign change in micelle mesophage. Inhibits bacterial flora of human saliva/gut at 0.25% as well as acting as a fungi static agent in aqueous dispersion (1%).

Solubility
Soluble in water (293 g/L).

Notes
Hygroscopic, store away form water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent and reducing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-16-6
Molecular Formula C15H28NNaO3
Molecular Weight (g/mol) 293.38
MDL Number MFCD00042728
InChI Key KSAVQLQVUXSOCR-UHFFFAOYSA-M
Synonym sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
PubChem CID 23668817
IUPAC Name sodium;2-[dodecanoyl(methyl)amino]acetate
SMILES [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

Specifications

Melting Point 44°C to 47°C
Color White
Linear Formula CH3(CH2)10CON(CH3)CH2COONa
Quantity 50 g
Beilstein 5322974
Sensitivity Hygroscopic
Merck Index 14,4368
Solubility Information Soluble in water (293g/L).
Formula Weight 293.38
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material N-Lauroylsarcosine sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.