missing translation for 'onlineSavingsMsg'
Learn More

N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine, 96%

Produktkod. 10337571 Handla allt< titel> Produkter
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
Packaging:
Glass bottle
Förpackningsstorlek:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Packaging Förpackningsstorlek
10337571 5 g Glass bottle 5g
15722166 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 10337571 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 344300050
 Restricted Item
In Stock
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Format
Denna artikel kan inte returneras. Se returpolicy

CAS: 93102-05-7 | C13H23NOSi | 237.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 93102-05-7
Molecular Formula C13H23NOSi
Molecular Weight (g/mol) 237.42
MDL Number MFCD00674005
InChI Key RPZAAFUKDPKTKP-UHFFFAOYSA-N
Synonym n-methoxymethyl-n-trimethylsilylmethyl benzylamine, n-benzyl-1-methoxy-n-trimethylsilyl methyl methanamine, benzyl methoxymethyl trimethylsilyl methyl amine, benzyl-methoxymethyl-trimethylsilanylmethyl-amine, n-benzyl-n-methoxymethyl-n-trimethylsilylmethylamine, n-benzyl-n-methoxymethyl trimethylsilylmethylamine, n-methoxymethyl phenyl-n-trimethylsilyl methyl methanamine, n-methoxymethyl-n-trimethylsilyl methyl phenyl, n-methoxymethyl-n-trimethylsilylmethyl-benzylamine
PubChem CID 353442
IUPAC Name N-(methoxymethyl)-1-phenyl-N-(trimethylsilylmethyl)methanamine
SMILES COCN(CC1=CC=CC=C1)C[Si](C)(C)C

Specifikationer

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9280g/mL
Elemental Analysis 95% min.
Boiling Point 76°C (0.3 mmHg)
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula C6H5CH2N(CH2OCH3)CH2Si(CH3)3
Refractive Index 1.4915 to 1.4935
Quantity 5 g
Specific Gravity 0.928
Formula Weight 237.42
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine
Signalord
  • Warning
Riskkategorier
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faroangivelser
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Skyddsangivelse
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.