missing translation for 'onlineSavingsMsg'
Learn More

n-methyl-[1-(2-furylmethyl)piperid-4-yl]methylamine, 97%, Thermo Scientific™

Product Code. 10225994
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10225994 1 g Amber Glass Bottle 1g
10153274 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10225994 Supplier Thermo Scientific Maybridge Supplier No. CC68546DA
 Restricted Item
Estimated Shipment: 08-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 934570-57-7
Molecular Formula C12H20N2O
Molecular Weight (g/mol) 208.31
MDL Number MFCD09966159
InChI Key LISJAFHYRUEIGV-UHFFFAOYSA-N
Synonym n-methyl-1-2-furylmethyl piperid-4-yl methylamine, n-1-2-furylmethyl piperidin-4-yl methyl-n-methylamine, 1-furan-2-ylmethyl piperidin-4-yl methyl methyl amine, 1-2-furylmethyl 4-piperidyl methyl methylamine, 1-furan-2-yl methyl piperidin-4-yl methyl methyl amine, 1-1-furan-2-ylmethyl piperidin-4-yl-n-methylmethanamine, 1-1-furan-2-yl methyl piperidin-4-yl-n-methylmethanamine
PubChem CID 42556092
SMILES CNCC1CCN(CC2=CC=CO2)CC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name ({1-[(furan-2-yl)methyl]piperidin-4-yl}methyl)(methyl)amine
Formula Weight 208.3
Percent Purity 97%
Chemical Name or Material N-Methyl-[1-(2-furylmethyl)piperid-4-yl]methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.