missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-1,3-propanediamine, 99%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
Beskrivelse
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | -72°C |
| Density | 0.8400g/mL |
| Boiling Point | 139°C to 141°C |
| Flash Point | 35°C |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass Bottle |
| Linear Formula | CH3NH(CH2)3NH2 |
| Refractive Index | 1.4468 |
| Vis mere |
Sikkerhed og håndtering
Produktidentifikator
- N-Methyl-1,3-propanediamine
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion