missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1,3-propanediamine, 99%

Product Code. 10784281 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
This item is not returnable. View return policy

Product Code. 10784281

Brand: Thermo Scientific Acros 155570010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolitik
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

CAS: 6291-84-5 | C4H12N2 | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 6291-84-5
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008209
InChI Key QHJABUZHRJTCAR-UHFFFAOYSA-N
Synonym n-methyl-1,3-propanediamine, n-methyl-1,3-diaminopropane, 3-methylamino propylamine, 1,3-propanediamine, n-methyl, n-methylpropane-1,3-diamine, 3-aminopropyl methylamine, n-methyltrimethylenediamine, 3-aminopropylmethylamine, n-methyl-1,3-propylenediamine, 1-amino-3-methylamino propane
PubChem CID 80511
IUPAC Name N'-methylpropane-1,3-diamine
SMILES CNCCCN

Tekniske data

CAS Min % 98.5
CAS Max % 100.0
Melting Point -72°C
Density 0.8400g/mL
Boiling Point 139°C to 141°C
Flash Point 35°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3NH(CH2)3NH2
Refractive Index 1.4468
Quantity 1 L
UN Number 2922
Beilstein 04,I,419
Specific Gravity 0.84
Solubility Information Solubility in water: soluble
Viscosity 1.6 mPa.s (20°C)
Formula Weight 88.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Methyl-1, 3-propanediamine, 99%
Vis mere Vis mindre
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • N-Methyl-1,3-propanediamine
Signalord
  • Danger
Farekategori
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Faresætning
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Sikkerhedssætning
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.