missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-[1-(6-methylpyrazin-2-yl)piperid-4-yl]methylamine, 90%, Thermo Scientific™

Product Code. 10225614
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10225614 1 g Amber Glass Bottle 1g
10215614 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10225614 Supplier Thermo Scientific Maybridge Supplier No. CC58946DA
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 887922-91-0
Molecular Formula C12H20N4
MDL Number MFCD09064987
Synonym n-methyl-1-6-methylpyrazin-2-yl piperid-4-yl methylamine, methyl 1-6-methylpyrazin-2-yl piperidin-4-yl methyl amine, methyl 1-6-methylpyrazin-2-yl 4-piperidyl methyl amine, n-methyl-1-1-6-methylpyrazin-2-yl piperidin-4-yl methanamine

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 220.32
Percent Purity 90%
Chemical Name or Material N-methyl-[1-(6-methylpyrazin-2-yl)piperid-4-yl]methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.