missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(1-methyl-1H-pyrazol-5-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10423214
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10423214 1 g Amber Glass Bottle 1g
10357314 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10423214 Supplier Thermo Scientific Maybridge Supplier No. CC62346DA
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 930111-04-9
Molecular Formula C6H11N3
Molecular Weight (g/mol) 125.18
MDL Number MFCD03419812
InChI Key CHGIPIYKGPNYKT-UHFFFAOYSA-N
Synonym n-methyl-1-methyl-1h-pyrazol-5-yl methylamine, methyl 1-methyl-1h-pyrazol-5-yl methyl amine, n-methyl 1-methyl-1h-pyrazol-5-yl methanamine, n,1-dimethyl-1h-pyrazole-5-methanamine, n-methyl-1-1-methyl-1h-pyrazol-5-yl methanamine, methyl 2-methylpyrazol-3-yl methyl amine, 1-methyl-5-methylamino methyl-1h-pyrazole, methyl-2-methyl-2h-pyrazol-3-ylmethyl-amine, methyl 1-methylpyrazol-5-yl methyl amine, 1-methyl-5-methylamino methyl pyrazole
PubChem CID 7019426
SMILES CNCC1=CC=NN1C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name N-methyl-1-(2-methylpyrazol-3-yl)methanamine
Formula Weight 125.17
Percent Purity 97%
Chemical Name or Material N-methyl-(1-methyl-1H-pyrazol-5-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.