missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1-naphthalenemethylamine hydrochloride, 98%

Product Code. 11472120
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472120 1 g 1g
11482120 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11472120 Supplier Thermo Scientific Alfa Aesar Supplier No. H28719.03
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 65473-13-4 | C12H14ClN | 207.701 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65473-13-4
Molecular Formula C12H14ClN
Molecular Weight (g/mol) 207.701
MDL Number MFCD00012555
InChI Key BVJVHPKFDIYQOU-UHFFFAOYSA-N
Synonym n-methyl-1-naphthalenemethylamine hydrochloride, n-methyl-1-naphthalen-1-yl methanamine hydrochloride, n-methyl-1-naphthylmethylamine hydrochloride, n-methyl-n-naphthylmethylamine hydrochloride, unii-z8vrt9b309, methyl naphthalen-1-ylmethyl amine hydrochloride, n-methyl-1-naphthalenemethanamine hydrochloride, n-methyl-c-naphthalen-1-yl methanamine hydrochloride, 1-naphthalenemethanamine, n-methyl-, hydrochloride 1:1, 1-methylaminomethyl naphthalene hydrochloride
PubChem CID 16211748
IUPAC Name N-methyl-1-naphthalen-1-ylmethanamine;hydrochloride
SMILES CNCC1=CC=CC2=CC=CC=C21.Cl

Specifications

Melting Point 190°C to 193°C
Quantity 1 g
Formula Weight 207.7
Percent Purity 98%
Chemical Name or Material N-Methyl-1-naphthalenemethylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.