missing translation for 'onlineSavingsMsg'
Learn More

N-methyl-(1H-benzimidazol-5-ylmethyl)amine dioxalate, 90%, Thermo Scientific™

Product Code. 11390627
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11390627 250 mg Amber Glass Bottle 250mg
10654405 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11390627 Supplier Thermo Scientific Maybridge Supplier No. CC45996CB
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 910036-85-0
Molecular Formula C13H15N3O8
Molecular Weight (g/mol) 341.276
MDL Number MFCD09025842
InChI Key HDQRBNFLWRHBAA-UHFFFAOYSA-N
Synonym n-methyl-1h-benzimidazol-5-ylmethyl amine dioxalate, 1h-1,3-benzodiazol-5-ylmethyl methyl amine; bis oxalic acid, n-methyl-1h-benzo d imidazol-5-ylmethyl amine dioxalate, oxalic acid-1-1h-benzimidazol-6-yl-n-methylmethanamine 2/1, 3h-1,3-benzodiazol-5-ylmethyl methyl amine; bis oxalic acid
PubChem CID 18525786
IUPAC Name 1-(3H-benzimidazol-5-yl)-N-methylmethanamine;oxalic acid
SMILES CNCC1=CC2=C(C=C1)N=CN2.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 341.27
Percent Purity 90%
Chemical Name or Material N-methyl-(1H-benzimidazol-5-ylmethyl)amine dioxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.