missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2-phenoxyethylamine, 98%

Product Code. 11469390
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469390 1 g 1g
11459390 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469390 Supplier Thermo Scientific Alfa Aesar Supplier No. H50258.03
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 37421-04-8 | C9H13NO | 151.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37421-04-8
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00085690
InChI Key GUMBNOITRNRLCS-UHFFFAOYSA-N
Synonym methyl 2-phenoxyethyl amine, n-methyl-2-phenoxyethylamine, methyl-2-phenoxy-ethyl-amine, n-methyl-2-phenoxy-ethylamin, ethylamine, n-methyl-2-phenoxy, ethanamine, n-methyl-2-phenoxy, n-methyl-n-2-phenoxyethyl amine, alpha-phenoxy-beta-methylaminoethane, acmc-1ae2y, n-methyl-2-phenoxyethylamin
PubChem CID 37732
IUPAC Name N-methyl-2-phenoxyethanamine
SMILES CNCCOC1=CC=CC=C1

Specifications

Linear Formula C6H5OCH2CH2NHCH3
Quantity 1 g
UN Number UN2735
Formula Weight 151.21
Percent Purity 98%
Chemical Name or Material N-Methyl-2-phenoxyethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.