missing translation for 'onlineSavingsMsg'
Learn More

n-methyl-(2-thien-2-ylpyrimidin-5-yl)methylamine, 90%, Thermo Scientific™

Product Code. 10706915
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10706915 1 g Amber Glass Bottle 1g
10431884 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10706915 Supplier Thermo Scientific Maybridge Supplier No. CC66546DA
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 921939-14-2
Molecular Formula C10H11N3S
Molecular Weight (g/mol) 205.279
MDL Number MFCD09879951
InChI Key VZHVJAYQGDCZAB-UHFFFAOYSA-N
Synonym n-methyl-2-thien-2-ylpyrimidin-5-yl methylamine, methyl 2-thiophen-2-yl pyrimidin-5-yl methyl amine, methyl 2-2-thienyl pyrimidin-5-yl methyl amine, n-methyl-1-2-thiophen-2-yl pyrimidin-5-yl methanamine, n-methyl-2-thiophen-2-ylpyrimidin-5-yl methylamine
PubChem CID 24229733
IUPAC Name N-methyl-1-(2-thiophen-2-ylpyrimidin-5-yl)methanamine
SMILES CNCC1=CN=C(N=C1)C2=CC=CS2

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.28
Percent Purity 90%
Chemical Name or Material N-methyl-(2-thien-2-ylpyrimidin-5-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.