missing translation for 'onlineSavingsMsg'
Learn More

n-methyl-(3-methoxythien-2-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10575754
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10575754 250 mg Amber Glass Bottle 250mg
10462114 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10575754 Supplier Thermo Scientific Maybridge Supplier No. CC68646CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 944450-93-5
Molecular Formula C7H11NOS
Molecular Weight (g/mol) 157.23
MDL Number MFCD11841072
InChI Key HDALHOAZSOWZLX-UHFFFAOYSA-N
Synonym 3-methoxythiophen-2-yl methyl methyl amine, 3-methoxythiophen-2-yl-n-methylmethylamine, n-methyl-3-methoxythien-2-yl methylamine, 3-methoxy-2-methylamino methyl thiophene, 1-3-methoxythiophen-2-yl-n-methylmethanamine, 3-methoxy 2-thienyl methyl methylamine
PubChem CID 43811050
IUPAC Name 1-(3-methoxythiophen-2-yl)-N-methylmethanamine
SMILES CNCC1=C(OC)C=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 157.24
Percent Purity 97%
Chemical Name or Material N-methyl-(3-methoxythien-2-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.