missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(3-methylisoxazol-5-ylmethyl)amine, 97%, Thermo Scientific™

Product Code. 10557684
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10557684 1 g Amber Glass Bottle 1g
10423204 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10557684 Supplier Thermo Scientific Maybridge Supplier No. CC27046DA
 Restricted Item
Estimated Shipment: 12-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 401647-22-1
Molecular Formula C6H10N2O
Molecular Weight (g/mol) 126.16
MDL Number MFCD08060065
InChI Key UQKWKRSUELRKSK-UHFFFAOYSA-N
Synonym n-methyl-1-3-methylisoxazol-5-yl methanamine, n-methyl-3-methylisoxazol-5-ylmethyl amine, methyl 3-methyl-1,2-oxazol-5-yl methyl amine, n-methyl 3-methylisoxazol-5-yl methanamine, methyl 3-methylisoxazol-5-yl methyl amine, n-methyl-1-3-methyl-1,2-oxazol-5-yl methanamine, 5-isoxazolemethanamine,n,3-dimethyl, 5-isoxazolemethanamine, n,3-dimethyl, methyl 3-methylisoxazol-5-ylmethyl amine, n-methyl-n-3-methylisoxazol-5-yl methyl amine
PubChem CID 25220779
IUPAC Name N-methyl-1-(3-methyl-1,2-oxazol-5-yl)methanamine
SMILES CNCC1=CC(C)=NO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 126.16
Percent Purity 97%
Chemical Name or Material N-methyl-(3-methylisoxazol-5-ylmethyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.