Learn More
N-Methyl-3-phenoxybenzylamine, 97%, Maybridge™
Brand: Maybridge CC57146DE
Code : 31
Additional Details : CAS Number : 129535-78-0 Weight : 0.05500kg Transport : UN number : 2735 Chem class : 8 Pack group : III
Quantity | 5g |
---|
Chemical Identifiers
129535-78-0 | |
213.28 | |
GDVLOOQWWBPGSV-UHFFFAOYSA-N | |
10512764 | |
CNCC1=CC(=CC=C1)OC2=CC=CC=C2 |
C14H15NO | |
MFCD02089413 | |
n-methyl-3-phenoxybenzylamine, n-methyl-1-3-phenoxyphenyl methanamine, methyl 3-phenoxyphenyl methyl amine, n-methyl-3-phenoxybenzenemethanamine, benzenemethanamine, n-methyl-3-phenoxy, acmc-209bgx, benzenemethanamine,n-methyl-3-phenoxy | |
N-methyl-1-(3-phenoxyphenyl)methanamine |
Specifications
N-Methyl-3-phenoxybenzylamine | |
129535-78-0 | |
100.0 | |
C14H15NO | |
n-methyl-3-phenoxybenzylamine, n-methyl-1-3-phenoxyphenyl methanamine, methyl 3-phenoxyphenyl methyl amine, n-methyl-3-phenoxybenzenemethanamine, benzenemethanamine, n-methyl-3-phenoxy, acmc-209bgx, benzenemethanamine,n-methyl-3-phenoxy | |
CNCC1=CC(=CC=C1)OC2=CC=CC=C2 | |
213.28 | |
213.28 |
5g | |
96.5 | |
97% | |
MFCD02089413 | |
GDVLOOQWWBPGSV-UHFFFAOYSA-N | |
N-methyl-1-(3-phenoxyphenyl)methanamine | |
10512764 | |
97% |