missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(3-phenylisoxazol-5-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10737875
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10737875 250 mg Amber Glass Bottle 250mg
10747875 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10737875 Supplier Thermo Scientific Maybridge Supplier No. CC64546CB
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 852431-00-6
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD09055377
InChI Key IUBUISGKALKRFN-UHFFFAOYSA-N
Synonym n-methyl-1-3-phenylisoxazol-5-yl methanamine, n-methyl 3-phenylisoxazol-5-yl methanamine, 5-isoxazolemethanamine,n-methyl-3-phenyl, n-methyl-3-phenylisoxazol-5-yl methylamine, methyl 3-phenyl-1,2-oxazol-5-yl methyl amine, n-methyl-1-3-phenyl-1,2-oxazol-5-yl methanamine, 5-methylamino methyl-3-phenylisoxazole, methyl-3-phenyl-isoxazol-5-ylmethyl-amine, methyl 3-phenyl-1,2-oxazol-5-yl methyl amine oxalate
PubChem CID 24229716
IUPAC Name N-methyl-1-(3-phenyl-1,2-oxazol-5-yl)methanamine
SMILES CNCC1=CC(=NO1)C2=CC=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 188.23
Percent Purity 97%
Chemical Name or Material N-methyl-(3-phenylisoxazol-5-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.