missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl(4-methylthien-2-yl)methylamine 97+%, Thermo Scientific™

Product Code. 10729225
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10729225 250 mg Amber Glass Bottle 250mg
10412644 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10729225 Supplier Thermo Scientific Maybridge Supplier No. CC30646CB
 Restricted Item
Estimated Shipment: 08-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kemiske identifikatorer

CAS 886851-27-0
Molecular Formula C7H11NS
Molecular Weight (g/mol) 141.23
MDL Number MFCD08435851
InChI Key JICZWIQRPDVYNI-UHFFFAOYSA-N
Synonym n-methyl 4-methylthien-2-yl methylamine, methyl 4-methylthiophen-2-yl methyl amine, 2-thiophenemethanamine,n,4-dimethyl, 2-thiophenemethanamine, n,4-dimethyl, methyl 4-methyl 2-thienyl methyl amine, n-methyl-1-4-methylthiophen-2-yl methanamine
PubChem CID 18525716
IUPAC Name N-methyl-1-(4-methylthiophen-2-yl)methanamine
SMILES CNCC1=CC(C)=CS1

Tekniske data

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 141.24
Percent Purity 97%
Chemical Name or Material N-Methyl(4-methylthien-2-yl)methylamine
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.