missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(4-thien-2-yltetrahydropyran-4-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10524654
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10524654 250 mg Amber Glass Bottle 250mg
10387333 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10524654 Supplier Thermo Scientific Maybridge Supplier No. CC62246CB
 Restricted Item
Estimated Shipment: 12-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 916790-87-9
Molecular Formula C11H17NOS
Molecular Weight (g/mol) 211.32
MDL Number MFCD09879934
InChI Key DBLNZQVTWSJQDY-UHFFFAOYSA-N
Synonym methyl 4-thiophen-2-yl oxan-4-yl methyl amine, n-methyl-4-thien-2-yltetrahydro-2h-pyran-4-yl methylamine, n-methyl-1-4-thiophen-2-yl oxan-4-yl methanamine, 4-methylamino methyl-4-thien-2-yltetrahydro-2h-pyran, n-methyl-4-thiophen-2-yltetrahydropyran-4-yl methylamine, n-methyl-1-4-thiophen-2-yl tetrahydro-2h-pyran-4-yl methanamine, methyl 4-2-thienyl 2h-3,4,5,6-tetrahydropyran-4-yl methyl amine
PubChem CID 24229671
SMILES CNCC1(CCOCC1)C1=CC=CS1

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name N-methyl-1-(4-thiophen-2-yloxan-4-yl)methanamine
Formula Weight 211.33
Percent Purity 90%
Chemical Name or Material N-methyl-(4-thien-2-yltetrahydropyran-4-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.