missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl[5-(2-furyl)pyrid-2-yl]methylamine 95%, Thermo Scientific™

Product Code. 10347504
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10347504 250 mg Amber Glass Bottle 250mg
10575574 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10347504 Supplier Thermo Scientific Maybridge Supplier No. CC55146CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 892502-04-4
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD10700054
InChI Key DOKOUBYFAJTDEI-UHFFFAOYSA-N
Synonym 5-furan-2-yl pyridin-2-yl methyl methyl amine, 2-pyridinemethanamine,5-2-furanyl-n-methyl, 5-fur-2-yl-2-methyl aminomethyl pyridine, 5-2-furyl 2-pyridyl methyl methylamine, 5-fur-2-yl pyridin-2-yl-n-methylmethylamine, 1-5-furan-2-yl pyridin-2-yl-n-methylmethanamine
PubChem CID 26343611
IUPAC Name 1-[5-(furan-2-yl)pyridin-2-yl]-N-methylmethanamine
SMILES CNCC1=NC=C(C=C1)C2=CC=CO2

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 188.23
Percent Purity 90%
Chemical Name or Material n-methyl[5-(2-furyl)pyrid-2-yl]methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.