missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(5-bromopyrid-3-yl)methylamine, 95%, Thermo Scientific™

Product Code. 10091194
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10091194 250 mg Amber Glass Bottle 250mg
10777675 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10091194 Supplier Thermo Scientific Maybridge Supplier No. CC67146CB
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 73335-64-5
Molecular Formula C7H9BrN2
Molecular Weight (g/mol) 201.07
MDL Number MFCD07375057
InChI Key UHDHQYPCHVXIKK-UHFFFAOYSA-N
Synonym 1-5-bromopyridin-3-yl-n-methylmethanamine, n-methyl-5-bromopyrid-3-yl methylamine, 5-bromopyridin-3-yl methyl methyl amine, n-methyl-5-bromopyridin-3-yl methylamine, 5-bromopyrid-3-ylmethyl methylamine, 3-bromo-5-methylamino methyl pyridine, 3-pyridinemethanamine,5-bromo-n-methyl, 5-bromo 3-pyridyl methyl methylamine, 5-bromopyridin-3-yl-n-methylmethylamine, 3-pyridinemethanamine, 5-bromo-n-methyl
PubChem CID 12515073
SMILES CNCC1=CC(Br)=CN=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 1-(5-bromopyridin-3-yl)-N-methylmethanamine
Formula Weight 201.07
Percent Purity 95%
Chemical Name or Material N-methyl-(5-bromopyrid-3-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.