missing translation for 'onlineSavingsMsg'
Learn More

n-methyl-(5-pyrid-3-ylthien-2-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10567294
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10567294 250 mg Amber Glass Bottle 250mg
10653265 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10567294 Supplier Thermo Scientific Maybridge Supplier No. CC66846CB
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 837376-49-5
Molecular Formula C11H12N2S
Molecular Weight (g/mol) 204.291
MDL Number MFCD11109328
InChI Key ZSTXLHXHAPANSL-UHFFFAOYSA-N
Synonym methyl 5-pyridin-3-yl thiophen-2-yl methyl amine, n-methyl-5-pyrid-3-ylthien-2-yl methylamine, n-methyl-5-pyridin-3-yl thiophen-2-yl methylamine, 2-methylamino methyl-5-pyridin-3-ylthiophene, n-methyl-1-5-pyridin-3-yl thiophen-2-yl methanamine, nicotine 3-heteroaromatic analogue 1b, methyl 5-3-pyridyl 2-thienyl methyl amine, 2-thiophenemethanamine,n-methyl-5-3-pyridinyl, 2-thiophenemethanamine, n-methyl-5-3-pyridinyl
PubChem CID 11321674
IUPAC Name N-methyl-1-(5-pyridin-3-ylthiophen-2-yl)methanamine
SMILES CNCC1=CC=C(S1)C2=CN=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 204.3
Percent Purity 97%
Chemical Name or Material N-methyl-(5-pyrid-3-ylthien-2-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.