missing translation for 'onlineSavingsMsg'
Learn More

N-methyl-bis(trifluoroacetamide), 98%

Product Code. 13488468 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass Bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13488468 25 mL Glass Bottle 25mL
10158742 5 mL Glass Bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13488468 Supplier Thermo Scientific Acros Supplier No. 210070250
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 685-27-8 | C5H3F6NO2 | 223.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 685-27-8
Molecular Formula C5H3F6NO2
Molecular Weight (g/mol) 223.07
InChI Key AWGBWLXGUPTXHF-UHFFFAOYSA-N
Synonym mbtfa, n-methyl-bis trifluoroacetamide, n-methylbis trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl acetamide, n,n-bis trifluoroacetyl methylamine, 2,2,2-trifluoro-n-methyl-n-2,2,2-trifluoroacetyl acetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl, n-methylbistrifluoroacetamide, n-methyl-bis-trifluoroacetamide
PubChem CID 69635
IUPAC Name 2,2,2-trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide
SMILES CN(C(=O)C(F)(F)F)C(=O)C(F)(F)F

Specifications

Density 1.5470g/mL
Boiling Point 120.0°C to 122.0°C
Flash Point 45°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula (CF3CO)2NCH3
Quantity 25 mL
Specific Gravity 1.547
Formula Weight 223.07
Percent Purity 98%
Chemical Name or Material N-Methyl-bis(trifluoroacetamide)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Methyl-bis(trifluoroacetamide)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.