missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(chroman-8-ylmethyl)amine, 97%, Thermo Scientific™

Product Code. 10737315
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10737315 1 g Amber Glass Bottle 1g
10652705 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10737315 Supplier Thermo Scientific Maybridge Supplier No. CC76146DA
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1048970-16-6
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD12198126
InChI Key VVCGFZWBAXPSIO-UHFFFAOYSA-N
Synonym n-methyl-chroman-8-ylmethyl amine, 3,4-dihydro-2h-1-benzopyran-8-ylmethyl methyl amine, chroman-8-ylmethyl methylamine, 1-3,4-dihydro-2h-1-benzopyran-8-yl-n-methylmethanamine
PubChem CID 43811059
IUPAC Name 1-(3,4-dihydro-2H-chromen-8-yl)-N-methylmethanamine
SMILES CNCC1=CC=CC2=C1OCCC2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 177.25
Percent Purity 97%
Chemical Name or Material N-methyl-(chroman-8-ylmethyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.