missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-D-aspartic acid, 98%

Product Code. p-160212 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10682382 50 mg Glass bottle 50mg
10239323 250 mg Glass bottle 250mg
10762515 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10682382 Supplier Thermo Scientific Acros Supplier No. 329190500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6384-92-5 | C5H9NO4 | 147.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6384-92-5
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.1
MDL Number MFCD00004226
InChI Key HOKKHZGPKSLGJE-GSVOUGTGSA-N
Synonym n-methyl-d-aspartic acid, nmda, n-methyl-d-aspartate, n-methylaspartate, d-aspartic acid, n-methyl, methyl aspartic acid, n-me-d-asp-oh, r-2-methylamino succinic acid, unii-1903b9q6pi, n methyl d aspartate
PubChem CID 22880
ChEBI CHEBI:31882
IUPAC Name (2R)-2-(methylamino)butanedioic acid
SMILES CNC(CC(=O)O)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 187°C to 192°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 50 mg
Merck Index 15, 6749
Specific Rotation Condition − 28.00 (20.00°C c=1,HCl 6N)
Specific Rotation -28
Solubility Information Solubility in water: slightly soluble in water. Other solubilities: soluble in dilute aqueous base,slightly soluble i
Formula Weight 147.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Methyl-D-aspartic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.