missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-D-cysteine Hydrochloride, TRC

Product Code. 30483375
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483375 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483375 Supplier Toronto Research Chemicals Supplier No. TRCM33854850MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Methyl-D-cysteine Hydrochloride
CAS 2519854-90-9
Molecular Formula C4H9NO2S . (HCl)
Quantity 50 mg
Synonym (2S)-2-(Methylamino)-3-sulfanylpropanoic Acid Hydrochloride
InChI Formula InChI=1S/C4H9NO2S.ClH/c1-5-3(2-8)4(6)7;/h3,5,8H,2H2,1H3,(H,6,7);1H/t3-;/m1./s1
SMILES SC[C@@H](N([H])C)C(O)=O.Cl
IUPAC Name methyl-D-cysteine hydrochloride
Molecular Weight (g/mol) 171.64
Formula Weight 171.02
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.