missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-D-glucamine, 100%, MP Biomedicals™

Product Code. 11414752 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414752 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11414752 Supplier MP Biomedicals Supplier No. 0219150691
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in the synthesis of surface active agents, pharmaceuticals and dyes.

Specifications

CAS 6284-40-8
Color White
Assay Percent Range 1
Molecular Formula C7H17NO5
Quantity 1 kg
Synonym meglumine, n-methyl-d-glucamine, n-methylglucamine, meglumin, 1-deoxy-1-methylamino-d-glucitol, methylglucamin, meglumina, 1-deoxy-1-methylaminosorbitol, d-glucitol, 1-deoxy-1-methylamino, n-methyl-d--glucamine
InChI Key MBBZMMPHUWSWHV-BDVNFPICSA-N
SMILES CNCC(C(C(C(CO)O)O)O)O
IUPAC Name (2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol
Molecular Weight (g/mol) 195.215
PubChem CID 8567
ChEBI CHEBI:59732
Formula Weight 195.22
Grade Not Available
Physical Form Powder
Chemical Name or Material N-Methyl-D-glucamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.